Learn More
Enzo Life Sciences CD38 Recombinant monoclonal antibody (HB7) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4370200
Encompass_Preferred
Clone: HB7. Isotype: IgGκ. Immunogen: Viable cells (1 x 10E7) of the human cell line, BJAB (Human Burkitt lymphoma B cell line), which was were injected intraperitoneally into 8-week-old BALB/c mice. Species reactivity: Human. Specificity: Recognizes an epitope between aa 273-285 of human CD38, an approximately 45 kDa type II transmembrane protein, expressed on essentially all pre-B lymphocytes, plasma cells, and thymocytes. Also present on activated T lymphocytes, natural killer (NK) lymphocytes, myeloblasts, and erythroblasts. Bimodally expressed during B cell development, modulating from high in immature cells to low in intermediate ones and back to high on mature B cells. This antibody competes with clone AT13/5. Applications: Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2964