Learn More
ACROBiosystems Recombinant Protein;Human CD38 Protein, Fc Tag;HEK293;1MG;CD8,CD38,T10,cADPr hydrolase 1
Supplier:  ACROBiosystems CD8H52551MG
Encompass_Preferred
Recombinant Protein;Human CD38 Protein, Fc Tag;1MG;Human CD38, Fc Tag (CD8-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # P28907-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.5 kDa. The protein migrates as 66-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD38, Fc Tag (Cat. No. CD8-H5255) at 2 μg/mL (100 μL/well) can bind Anti-CD38, Human IgG1 with a linear range of 0.6-2 ng/mL (QC tested).
Catalog No. 50-165-8197