Learn More
ACROBiosystems Recombinant Protein;Cynomolgus CD38 Protein;HEK293;1MG;CD8,CD38,T10,cADPr hydrolase 1
Supplier:  ACROBiosystems CD8C52231MG
Encompass_Preferred
Recombinant Protein;Cynomolgus CD38 Protein;1MG;Cynomolgus CD38 (CD8-C5223) is expressed from human 293 cells (HEK293). It contains AA Leu 44 - Ile 301 (Accession # Q5VAN0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.8 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7657