Learn More
CAPRICO BIOTECHNOLOGIES INC CD39 FITC Antibody, clone BU61
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4084015
Encompass_Preferred
The CD39-FITC Antibody from Caprico Biotechnologies is a fluorescein isothiocyanate (FITC) conjugated monoclonal antibody that recognizes human CD39. CD39 is an ecto-apyrase involved in suppressing immune responses by hydrolyzing ATP and ADP to AMP. This antibody can be used for flow cytometry, immunofluorescence microscopy, and other fluorescent detection methods. It is supplied as a liquid in PBS with 0.1% sodium azide and 1% BSA. The immunogen is a recombinant human CD39 protein.
Catalog No. 50-227-9144