Learn More
Enzo Life Sciences CD39 (human) monoclonal antibody (AC2.5) (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2370100
Encompass_Preferred
Clone: AC2.5. Isotype: IgG1. Immunogen: EBV-transformed human B lymphoblastoid cell line. Species reactivity: Human. Applications: IHC, WB. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2784