Learn More
CAPRICO BIOTECHNOLOGIES INC CD39 PE-Cyanine7 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4084085
Encompass_Preferred
The CD39 PE-Cyanine7 Antibody is a mouse monoclonal antibody that binds to human CD39. It is conjugated to PE-Cyanine7 and is suitable for flow cytometry applications. This antibody recognizes the ecto-ATPase CD39, which is expressed on regulatory T cells and other immune cell types. It can be used to identify and characterize CD39+ cell populations by flow cytometry. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each test contains 100 μL of antibody.
Catalog No. 50-227-9145