Learn More
Enzo Life Sciences CD39L1 (mouse) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX215044R100
Encompass_Preferred
Host: Rabbit. Immunogen: Vector containing the cDNA of mouse CD39L1. UniProt ID: O55026. Species reactivity: Mouse. Applications: Flow Cytometry, ICC. Recommended Dilutions: Flow Cytometry (1:100-1:400) Immunocytochemistry (1:100-1:400) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Antiserum diluted with PBS containing 0.1% BSA and 0.02% sodium azide.
Catalog No. 50-200-8206