Learn More
ACROBiosystems Recombinant Protein;Cynomolgus CD3 delta Protein;HEK293;1MG;CDD,CD3D,CD3-DELTA,T3D,CD3 DELTA
Catalog No. 501659125
Encompass_Preferred
Recombinant Protein;Cynomolgus CD3 delta Protein;1MG;Cynomolgus CD3D, His Tag (CDD-C52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession # Q95LI8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.3 kDa. The protein migrates as 26-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9125 Supplier ACROBiosystems Supplier No. CDDC52H81MG
Recombinant Protein;Cynomolgus CD3 delta Protein;1MG;Cynomolgus CD3D, His Tag (CDD-C52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession # Q95LI8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.3 kDa. The protein migrates as 26-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.