Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 916MSM8P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: C3e/1308. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: Jurkat cells. Tonsil and lymph node. Cellular Staining: Cell surface and cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CD3e fragment (aa 23-119). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 20kDa.Chromosome: 11q23.3. Gene Symbol: CD3E. Gene ID: 916. Ensemble: ENSG00000198851. Uniprot: NP_000724.
Catalog No. 50-188-5322