Learn More
Aviva Systems Biology Corp CD3E Antibody - middle region (ARP59097_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59097_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD3E. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CD3E. Alternate names: FLJ18683, T3E, TCREApplicationWB. Species reactivity: Human
Catalog No. 50-286-577