Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag™&Tag Free
Catalog No. 501956833
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5-1 (CD3E) & Q95LI8-1 (CD3D)).;Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free is produced by co-expression of CD3E and CD3D, has a calculated MW of 17.8 kDa (CD3E) and 13.5 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag and followed by an Avi tag (Avitag™) and subunit CD3D contains no tag at the C-terminus. The reducing (R) heterodimer protein migrates as 21-23 kDa (CD3E) and 19-21 kDa (CD3D) due to glyosylation.;Immobilized Mouse Anti-Human CD3 (SP34-2) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free (Cat. No. CDD-C82W6) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-195-6833 Supplier ACROBiosystems Supplier No. CDDC82W6200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5-1 (CD3E) & Q95LI8-1 (CD3D)).;Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free is produced by co-expression of CD3E and CD3D, has a calculated MW of 17.8 kDa (CD3E) and 13.5 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag and followed by an Avi tag (Avitag™) and subunit CD3D contains no tag at the C-terminus. The reducing (R) heterodimer protein migrates as 21-23 kDa (CD3E) and 19-21 kDa (CD3D) due to glyosylation.;Immobilized Mouse Anti-Human CD3 (SP34-2) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free (Cat. No. CDD-C82W6) with a linear range of 20-156 ng/mL (QC tested).