Learn More
ACROBiosystems ACROBiosystems Cynomolgus CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin
Catalog No. 501956965
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin (CDD-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5-1 (CD3E) & Q95LI8-1 (CD3D)).;Cynomolgus CD3E & CD3D Heterodimer Protein, Llama IgG2b Fc Tag & Llama IgG2b Fc Tag, low endotoxin is produced by co-expression of CD3E and CD3D, has a calculated MW of 41.9 kDa (CD3E) and 40.8 kDa (CD3D). Subunit CD3E is fused with a llama IgG2b Fc tag at the C-terminus and subunit CD3D is fused with a llama IgG2b Fc tag at the C-terminus. The predicted N-terminus is Gln 22 (CD3E) & Phe 22 (CD3D). The reducing (R) protein migrates as 50 kDa and 55-65 kDa due to glycosylation.;Immobilized Cynomolgus CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin (Cat. No. CDD-C5259) at 2 μg/mL (100 μL/well) can bind Mouse Anti-Human CD3 (SP34-2) (mouse IgG1) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-195-6965 Supplier ACROBiosystems Supplier No. CDDC5259100UG
Recombinant Protein;100UG;Cynomolgus CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin (CDD-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5-1 (CD3E) & Q95LI8-1 (CD3D)).;Cynomolgus CD3E & CD3D Heterodimer Protein, Llama IgG2b Fc Tag & Llama IgG2b Fc Tag, low endotoxin is produced by co-expression of CD3E and CD3D, has a calculated MW of 41.9 kDa (CD3E) and 40.8 kDa (CD3D). Subunit CD3E is fused with a llama IgG2b Fc tag at the C-terminus and subunit CD3D is fused with a llama IgG2b Fc tag at the C-terminus. The predicted N-terminus is Gln 22 (CD3E) & Phe 22 (CD3D). The reducing (R) protein migrates as 50 kDa and 55-65 kDa due to glycosylation.;Immobilized Cynomolgus CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin (Cat. No. CDD-C5259) at 2 μg/mL (100 μL/well) can bind Mouse Anti-Human CD3 (SP34-2) (mouse IgG1) with a linear range of 1-20 ng/mL (QC tested).