Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD3E & CD3D, ultra sensitivity (primary amine labeling);HEK293;200UG;CDD,CD3E & CD3D,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3D,CD3-DELTA,T3D,CD3E&CD3D,CD3 delta & CD3 epsilon,CD3 delta / epsilon
Catalog No. 501659181
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD3E & CD3D, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human CD3 epsilon & CD3 delta (CDD-H82W0) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # NP_000724.1 (CD3E) & AAH39035 (CD3D)). It is the biotinylated form of Human CD3 epsilon & CD3 delta Protein (Cat # CDD-H52W0).;The subunit CD3E carries a human IgG1 Fc fragment at the C-terminus, followed by a polyhistidine tag.The subunit CD3D carries a human IgG1 Fc fragment at the C-terminus, followed by a flag tag. The protein has a calculated MW of 39.2 kDa (CD3E), 37.5 kDa (CD3D) . The protein migrates as 40-55 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Biotinylated Human CD3E & CD3D (Cat. No. CDD-H82W0) can bind OKT3 with a linear range of 2-39 ng/mL. The biotinylated CD3E & CD3D heterodimers are coated at 1 ug/mL (100 L/well) on a streptavidin c
Catalog No. 50-165-9181 Supplier ACROBiosystems Supplier No. CDDH82W0200UG
Recombinant Protein;Biotinylated Human CD3E & CD3D, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human CD3 epsilon & CD3 delta (CDD-H82W0) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # NP_000724.1 (CD3E) & AAH39035 (CD3D)). It is the biotinylated form of Human CD3 epsilon & CD3 delta Protein (Cat # CDD-H52W0).;The subunit CD3E carries a human IgG1 Fc fragment at the C-terminus, followed by a polyhistidine tag.The subunit CD3D carries a human IgG1 Fc fragment at the C-terminus, followed by a flag tag. The protein has a calculated MW of 39.2 kDa (CD3E), 37.5 kDa (CD3D) . The protein migrates as 40-55 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Biotinylated Human CD3E & CD3D (Cat. No. CDD-H82W0) can bind OKT3 with a linear range of 2-39 ng/mL. The biotinylated CD3E & CD3D heterodimers are coated at 1 ug/mL (100 L/well) on a streptavidin c