Learn More
ACROBiosystems Recombinant Protein;Cynomolgus CD3 epsilon & CD3 delta Heterodimer Protein, His Tag & Flag Tag;HEK293;1MG;CDD,CD3E & CD3D,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3D,CD3-DELTA,T3D,CD3E&CD3D,CD3 delta & CD3 epsilon,CD3 delta / epsilon
Catalog No. 501659123
Encompass_Preferred
Recombinant Protein;Cynomolgus CD3 epsilon & CD3 delta Heterodimer Protein, His Tag & Flag Tag;1MG;Recombinant Cynomolgus CD3E & CD3D heterodimer protein (Cynomolgus CD3E & CD3D heterodimer) Gln 22 - Asp 117 (CD3E,Q95LI5) & Phe 22 - Ala 105 (CD3D,Q95LI8) was produced in human 293 cells (HEK293) at ACROBiosystems.;The heterodimer Cynomolgus CD3E & CD3D heterodimer is produced by co-expression of CD3E and CD3D, has a calculated MW of 15.5 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with his tag at the C-terminus and subunit CD3D is fused with flag tag at the C-terminus. The predicted N-terminus is Gln 22 (CD3E) and Phe 22 (CD3D). The reducing (R) heterodimer protein migrates as 19-22 kDa and 25-40 kDa due to glyosylation.;Immobilized SP34-2 at 5 μg/mL (100 μL/well) can bind Cynomolgus CD3E & CD3D, His Tag, Flag Tag (Cat. No. CDD-C52W4) with a linear range of 0.6-5 ng/mL (Routinely tested).
Catalog No. 50-165-9123 Supplier ACROBiosystems Supplier No. CDDC52W41MG
Recombinant Protein;Cynomolgus CD3 epsilon & CD3 delta Heterodimer Protein, His Tag & Flag Tag;1MG;Recombinant Cynomolgus CD3E & CD3D heterodimer protein (Cynomolgus CD3E & CD3D heterodimer) Gln 22 - Asp 117 (CD3E,Q95LI5) & Phe 22 - Ala 105 (CD3D,Q95LI8) was produced in human 293 cells (HEK293) at ACROBiosystems.;The heterodimer Cynomolgus CD3E & CD3D heterodimer is produced by co-expression of CD3E and CD3D, has a calculated MW of 15.5 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with his tag at the C-terminus and subunit CD3D is fused with flag tag at the C-terminus. The predicted N-terminus is Gln 22 (CD3E) and Phe 22 (CD3D). The reducing (R) heterodimer protein migrates as 19-22 kDa and 25-40 kDa due to glyosylation.;Immobilized SP34-2 at 5 μg/mL (100 μL/well) can bind Cynomolgus CD3E & CD3D, His Tag, Flag Tag (Cat. No. CDD-C52W4) with a linear range of 0.6-5 ng/mL (Routinely tested).