Learn More
ACROBiosystems ACROBiosystems Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free
Catalog No. 501956379
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free (CDG-C52W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Gln 23 - Thr 113 (CD3G) (Accession # Q95LI5-1 (CD3E) & Q95LI7-1 (CD3G)).;Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.0 kDa (CD3E) and 14.4 kDa (CD3G). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3G contains no tag at the C-terminus.
Catalog No. 50-195-6379 Supplier ACROBiosystems Supplier No. CDGC52W61MG
Recombinant Protein;1MG;Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free (CDG-C52W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Gln 23 - Thr 113 (CD3G) (Accession # Q95LI5-1 (CD3E) & Q95LI7-1 (CD3G)).;Cynomolgus CD3E&CD3G Heterodimer Protein, His Tag&Tag Free is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.0 kDa (CD3E) and 14.4 kDa (CD3G). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3G contains no tag at the C-terminus.