Learn More
Enzo Life Sciences CD3 epsilon Recombinant monoclonal antibody (12F6) (Mouse IgG2a kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6210200
Encompass_Preferred
Clone: 12F6. Isotype: IgG2aκ. Immunogen: Human CD3. Species reactivity: Human. Applications: Flow Cytometry. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3145