Learn More
Enzo Life Sciences CD3 epsilon Recombinant monoclonal antibody (500A2) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4270200
Encompass_Preferred
Clone: 500A2. Isotype: IgGκ. Immunogen: T cell receptor complexes derived from mouse C6VL-BS thymoma cells (C57BL/Ka). Species reactivity: Mouse. Specificity: Recognizes the 25-kDa ε chain of the T-cell receptor-associated CD3 complex expressed on thymocytes, mature T lymphocytes, and NK-T cells on many mouse strains. Applications: Flow Cytometry. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2954