Learn More
ACROBiosystems Mouse CD3 Epsilon Protein, Fc Tag, 1 mg
Catalog No. 501957324
Encompass_Preferred
Mouse CD3 epsilon, also known as CD3E, is a T-cell surface single-pass type I membrane glycoprotein. Together with other subunits and the T-cell receptor, it forms the T cell receptor-CD3 complex, which is essential for coupling antigen recognition to intracellular signal-transduction pathways and T-cell development.
  • Expressed from HEK293 cells with a human IgG1 Fc tag at the C-terminus.
  • Purity of >95% as determined by SDS-PAGE.
  • Endotoxin level is <1.0 EU per μg by the LAL method.
  • Calculated molecular weight of 36 kDa, migrating as 40–43 kDa under reducing conditions due to glycosylation.
  • Lyophilized from a 0.22 μm filtered solution in Tris with Glycine, Arginine, and NaCl, pH 7.5 with trehalose as a protectant.
  • Recommended for storage at -20°C or lower in lyophilized state.
Catalog No. 50-195-7324 Supplier ACROBiosystems Supplier No. CDEM52561MG
Mouse CD3 epsilon, also known as CD3E, is a T-cell surface single-pass type I membrane glycoprotein. Together with other subunits and the T-cell receptor, it forms the T cell receptor-CD3 complex, which is essential for coupling antigen recognition to intracellular signal-transduction pathways and T-cell development.
  • Expressed from HEK293 cells with a human IgG1 Fc tag at the C-terminus.
  • Purity of >95% as determined by SDS-PAGE.
  • Endotoxin level is <1.0 EU per μg by the LAL method.
  • Calculated molecular weight of 36 kDa, migrating as 40–43 kDa under reducing conditions due to glycosylation.
  • Lyophilized from a 0.22 μm filtered solution in Tris with Glycine, Arginine, and NaCl, pH 7.5 with trehalose as a protectant.
  • Recommended for storage at -20°C or lower in lyophilized state.