Learn More
ACROBiosystems ACROBiosystems Cynomolgus CD3 epsilon Protein, Fc,His Tag
Catalog No. 501957530
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus CD3 epsilon, Fc,His Tag (CDE-C5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession # Q95LI5).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 38.3 kDa. The protein migrates as 43-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SP34-2 at 2 μg/mL (100 μL/well) can bind Cynomolgus CD3 epsilon, Fc,His Tag (Cat. No. CDE-C5254) with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-195-7530 Supplier ACROBiosystems Supplier No. CDEC5254100UG
Recombinant Protein;100UG;Cynomolgus CD3 epsilon, Fc,His Tag (CDE-C5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession # Q95LI5).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 38.3 kDa. The protein migrates as 43-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SP34-2 at 2 μg/mL (100 μL/well) can bind Cynomolgus CD3 epsilon, Fc,His Tag (Cat. No. CDE-C5254) with a linear range of 0.2-3 ng/mL (Routinely tested).