Learn More
ACROBiosystems Recombinant Protein;Human CD3 epsilon Protein;HEK293;1MG;CDE,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon ,CDE-H822R
Catalog No. 501658950
Encompass_Preferred
Recombinant Protein;Human CD3 epsilon Protein;1MG;Human CD3E, His Tag (CDE-H5223) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.2 kDa. The protein migrates as 15-16 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Anti Human CD3 MAb, mouse IgG1 (Clone # SP34-2) at 2 μg/mL (100 μL/well) can bind Human CD3E, His Tag (Cat. No. CDE-H5223) with a linear range of 0.4-6.25 ng/mL (QC tested).
Catalog No. 50-165-8950 Supplier ACROBiosystems Supplier No. CDEH52231MG
Recombinant Protein;Human CD3 epsilon Protein;1MG;Human CD3E, His Tag (CDE-H5223) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.2 kDa. The protein migrates as 15-16 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Anti Human CD3 MAb, mouse IgG1 (Clone # SP34-2) at 2 μg/mL (100 μL/well) can bind Human CD3E, His Tag (Cat. No. CDE-H5223) with a linear range of 0.4-6.25 ng/mL (QC tested).