Learn More
ACROBiosystems Recombinant Protein;Cynomolgus CD3 epsilon Protein;HEK293;1MG;CDE,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon ,CDE-H822R
Supplier:  ACROBiosystems CDEC52261MG
Encompass_Preferred
Recombinant Protein;Cynomolgus CD3 epsilon Protein;1MG;Cynomolgus CD3 epsilon (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession # Q95LI5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.2 kDa. The protein migrates as 10 kDa and 14-21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti Human CD3E MAb at 2 μg/mL (100 μL/well) can bind Cynomolgus CD3 epsilon (Cat. No. CDE-C5226) with a linear range of 0.05-1.6 ng/mL (QC tested).
Catalog No. 50-165-7438