Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD3 epsilon Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;CDE,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon ,CDE-H822R
Catalog No. 501657615
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD3 epsilon Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CD3 epsilon, His Tag, Avi Tag (CDE-H82E1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # P07766-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 15.5 kDa. The protein migrates as 17-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti Human CD3e MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD3 epsilon, His Tag, Avi Tag (Cat. No. CDE-H82E1) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-165-7615 Supplier ACROBiosystems Supplier No. CDEH82E1200UG
Recombinant Protein;Biotinylated Human CD3 epsilon Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CD3 epsilon, His Tag, Avi Tag (CDE-H82E1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # P07766-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 15.5 kDa. The protein migrates as 17-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti Human CD3e MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD3 epsilon, His Tag, Avi Tag (Cat. No. CDE-H82E1) with a linear range of 0.2-6 ng/mL (QC tested).