Learn More
Biogenex Laboratories CD4 : 4B12 : 6mL
SDP
Supplier:  Biogenex Laboratories AM4215M
Encompass_Preferred
CD4 is a 55-60 kD cell-surface glycoprotein, which participates in the molecular complexes involved in both T cell development and its antigen recognizing activity, by binding to the nonpolymorphic region of class II MHC. CD4 is considered as a stage marker of T cell development in the thymus, for it is expressed on the cell surface in a stage specific manner, during T cell development. This antibody reacts on a low level with human monocytes and macrophages but does not react with B-cells , granulocytes and thrombocytes.
Catalog No. 50-197-8767