Learn More
Aviva Systems Biology Corp CD4 Antibody - C-terminal region (ARP59105_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59105_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CD4. Alternate names: CD4mutApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-580