Learn More
Sino Biological CD4 Antibody (APC), Mouse MAb 25Tests
Supplier:  Sino Biological 60003MM02A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ferret CD4 (rFerret CD4; Catalog#60003-F08H; ABS50090.1; Met1-Leu401) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-224-8082