Learn More
Sino Biological CD4 Antibody (PE), Mouse MAb 25Tests
Supplier:  Sino Biological 60003MM02P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ferret CD4 (rF CD4; Catalog#60003-F08H; ABS50090.1; Met 1-Leu 401) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-7719