Learn More
CAPRICO BIOTECHNOLOGIES INC CD4 Antibody, 4120045
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4120045
Encompass_Preferred
The Caprico CD4 Antibody is a mouse monoclonal antibody that recognizes human CD4. It is produced in ascites and purified by protein A/G chromatography. This antibody can be used for ELISA, Western blot, immunoprecipitation, and flow cytometry applications. It detects CD4 protein expression on the surface of helper T cells. The immunogen is a recombinant protein corresponding to amino acids 1-178 of human CD4. This antibody is validated for flow cytometry analysis of CD4 expression on human peripheral blood lymphocytes.
Catalog No. 50-227-9147