Learn More
CAPRICO BIOTECHNOLOGIES INC CD4 APC-Cyanine7 Antibody Clone OKT4, 100 tests
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4120095
Encompass_Preferred
The CD4 APC Cyanine7 Antibody from Caprico Biotechnologies is a mouse monoclonal antibody that binds to human CD4. It is conjugated to APC-Cyanine7 and is recommended for flow cytometry applications. This antibody recognizes the D3 domain of CD4 and has been tested on human peripheral blood samples. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each test contains 100 μL of antibody at a concentration of 20 μg/mL.
Catalog No. 50-227-9150