Learn More
Cayman Chemical CD4 EXrAClular DomaInhum 100ug
SDP
Catalog No. 502948607
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-tagged CD4 expressed in HEK293 cells • Amino acids: 26-390 • MW: 67.4 kDa
Catalog No. 50-294-8607 Supplier Cayman Chemical Supplier No. 31831100 UG
Source: Active recombinant C-terminal human IgG1 Fc-tagged CD4 expressed in HEK293 cells • Amino acids: 26-390 • MW: 67.4 kDa