Learn More
Cayman Chemical CD4 Extracelular DomaInhum 1mg
SDP
Supplier:  Cayman Chemical 318311 MG
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-tagged CD4 expressed in HEK293 cells • Amino acids: 26-390 • MW: 67.4 kDa
Catalog No. 50-294-8933