Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human CD4 Protein, Fc Tag
Catalog No. 501956622
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CD4, Fc Tag (CD4-HF255) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730). It is the FITC labeled form of Human CD4, Fc Tag (CD4-H5259).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.5 kDa. The protein migrates as 75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human CD4, Fc Tag (Cat. No. CD4-HF255) with a linear range of 0.039-0.313 μg/mL (QC tested).
Catalog No. 50-195-6622 Supplier ACROBiosystems Supplier No. CD4HF255200UG
Recombinant Protein;200UG;FITC-Labeled Human CD4, Fc Tag (CD4-HF255) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730). It is the FITC labeled form of Human CD4, Fc Tag (CD4-H5259).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.5 kDa. The protein migrates as 75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human CD4, Fc Tag (Cat. No. CD4-HF255) with a linear range of 0.039-0.313 μg/mL (QC tested).