Learn More
ACROBiosystems ACROBiosystems Rhesus macaque CD4 Protein, Fc Tag
Catalog No. 501957214
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque CD4, Fc Tag (CD4-C5256) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # G7N5T8).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.0 kDa. The protein migrates as 67-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) at 10 μg/mL (100 μL/well) can bind Rhesus macaque CD4, Fc Tag (Cat. No. CD4-C5256) with a linear range of 0.04-0.31 μg/mL (QC tested).
Catalog No. 50-195-7214 Supplier ACROBiosystems Supplier No. CD4C52561MG
Recombinant Protein;1MG;Rhesus macaque CD4, Fc Tag (CD4-C5256) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # G7N5T8).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.0 kDa. The protein migrates as 67-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) at 10 μg/mL (100 μL/well) can bind Rhesus macaque CD4, Fc Tag (Cat. No. CD4-C5256) with a linear range of 0.04-0.31 μg/mL (QC tested).