Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human CD4 Protein, His Tag
Supplier:  ACROBiosystems CD4HF2H7200UG
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # AAH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.5 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human CD4, His Tag (Cat. No. CD4-HF2H7) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-195-6618