Learn More
Enzo Life Sciences CD4 (human) monoclonal antibody (MEM-115) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805053C100
Encompass_Preferred
Clone: MEM-115. Isotype: IgG2a. Immunogen: Human thymocytes and T lymphocytes. Species reactivity: Human. Specificity: Recognizes an epitope in the D1 domain of human CD4. Applications: Flow Cytometry, IP, FUNC. Purity: Protein A-affinity purified from ascites. Formulation: Liquid. In TBS, pH 8.0, containing 15mM sodium azide.
Catalog No. 50-201-0047