Learn More
Medchemexpress LLC CD4 Human HEK293 5ug
SDP
Catalog No. 5000183090
Encompass_Preferred
The CD4 protein is an integral membrane glycoprotein that plays a key role in immune responses In T cells it serves as a coreceptor for MHC class II molecules interacting with T cell receptors and initiating intracellular signaling pathways CD4 Protein Human (HEK293 Fc) is the recombinant human-derived CD4 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-83090 Supplier Medchemexpress LLC Supplier No. HYP729025UG
The CD4 protein is an integral membrane glycoprotein that plays a key role in immune responses In T cells it serves as a coreceptor for MHC class II molecules interacting with T cell receptors and initiating intracellular signaling pathways CD4 Protein Human (HEK293 Fc) is the recombinant human-derived CD4 protein expressed by HEK293 with C-hFc labeled tag