Learn More
CAPRICO BIOTECHNOLOGIES INC CD4 iFluor 700 Antibody, 4004195
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4004195
Encompass_Preferred
The Caprico CD4 iFluor 700 Antibody is a mouse monoclonal antibody that specifically binds to human CD4. It is conjugated to iFluor 700 and is suitable for flow cytometry, immunofluorescence microscopy, and immunohistochemistry. This antibody recognizes the D3 domain of CD4 and does not cross-react with other human antigens. It can be used to identify CD4+ T helper cells and is recommended for use in cell sorting. The antibody is available in multiple sizes ranging from 25 tests to 1000 tests.
Catalog No. 50-227-9159