Learn More
Genscript Corporation CD4/LEU3 His Rhesus macaque
Catalog No. 5000056853
Encompass
CD4 also known as L3T4 T4 and W3/25 is an approximately 55 kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T lymphocytes It is a standard phenotype marker for the identification of T cell populations Integral membrane glycoprotein that plays an essential role in the immune response and serves multiple functions in responses against both external and internal offenses In T-cells functions primarily as a coreceptor for MHC class II molecule peptide complex
Catalog No. 50-000-56853 Supplier Genscript Corporation Supplier No. Z04017100
CD4 also known as L3T4 T4 and W3/25 is an approximately 55 kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T lymphocytes It is a standard phenotype marker for the identification of T cell populations Integral membrane glycoprotein that plays an essential role in the immune response and serves multiple functions in responses against both external and internal offenses In T-cells functions primarily as a coreceptor for MHC class II molecule peptide complex