Learn More
Sino Biological Recombinant Human CD4 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10400H02H100
Encompass_Preferred
A DNA sequence encoding the human CD4 (NP_000607.1) (Met1-Trp390) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-4905