Learn More
Sino Biological Recombinant Human CD4 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10400H08HB100
Encompass_Preferred
A DNA sequence encoding the human CD4 (NP_000607.1) extracellular domain (Met1-Trp390) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6149