Learn More
Medchemexpress LLC CD4 Rat HEK293 Hi 10ug
SDP
Catalog No. 5000181645
Encompass_Preferred
CD4 Protein is expressed on the surface of helper T cells It plays a key role in immune responses by binding to major histocompatibility complex class II molecules on antigen-presenting cells facilitating T cell activation and cytokine production CD4 Protein is also involved in immune regulation and tolerance Understanding its functions can aid in developing therapies for autoimmune diseases and immunodeficiencies CD4 Protein Rat (HEK293 His) is the recombinant rat-derived CD4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81645 Supplier Medchemexpress LLC Supplier No. HYP7290510UG
CD4 Protein is expressed on the surface of helper T cells It plays a key role in immune responses by binding to major histocompatibility complex class II molecules on antigen-presenting cells facilitating T cell activation and cytokine production CD4 Protein is also involved in immune regulation and tolerance Understanding its functions can aid in developing therapies for autoimmune diseases and immunodeficiencies CD4 Protein Rat (HEK293 His) is the recombinant rat-derived CD4 protein expressed by HEK293 with C-His labeled tag