Learn More
Enzo Life Sciences CD4 Recombinant monoclonal antibody (13B8.2) (Mouse IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4420200
Encompass_Preferred
Clone: 13B8.2. Isotype: IgG1κ. Immunogen: Human HSB2-T cells. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IHC (FS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2969