Learn More
Enzo Life Sciences CD4 Recombinant monoclonal antibody (CE9.1) (Human IgG4 lambda) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4400200
Encompass_Preferred
Clone: CE9.1 (Clenoliximab). Isotype: IgG4λ. Immunogen: Domain 1 of human CD4. Species reactivity: Human Chimpanzee. Specificity: The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 µg/ml. Applications: ELISA, Flow Cytometry, IHC, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2967