Learn More
Enzo Life Sciences CD4 Recombinant monoclonal antibody (MT310) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4430200
Encompass_Preferred
Clone: MT310. Isotype: IgGκ. Species reactivity: Human. Specificity: Recognizes the extracellular domain D1 of CD4 – the same site as the HIV-1 gp120-binding site. Applications: Flow Cytometry, IF, IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2970