Learn More
Enzo Life Sciences CD4 Recombinant monoclonal antibody (YNB46.1.8) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4300200
Encompass_Preferred
Clone: YNB46.1.8 (Campath-9H). Isotype: IgGκ. Immunogen: Rat T-cell line (NB2) transfected with the human CD4. Species reactivity: Human. Specificity: Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes. Applications: Flow Cytometry, IHC (FS). Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2957