Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 958MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: C40/1605. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: L-428 Cells. Tonsil, Spleen or Thymus. Cellular Staining: Cell Surface and Secreted. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length CD40 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 43kDa.Chromosome: 20q12-q13.2. Gene Symbol: CD40. Gene ID: 958. Ensemble: ENSG00000101017. Uniprot: XP_016883624.
Catalog No. 50-188-5354