Learn More
Aviva Systems Biology Corp CD40 Antibody - N-terminal region (ARP41928_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41928_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD40. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CD40. Alternate names: Bp50, CDW40, MGC9013, TNFRSF5, p50ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-58049