Learn More
Sino Biological CD40 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 90097T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus CD40 ( Catalog#90097-C08H; NP_001252791.1; Met1-Arg193). CD40 specific IgG was purified by Rhesus CD40 affinity chromatography.
Catalog No. 50-224-6556