Learn More
ACROBiosystems ACROBiosystems Human CD40 / TNFRSF5 Protein, Fc Tag (HPLC-verified)
Supplier:  ACROBiosystems CD0H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD40, Fc Tag (HPLC-verified) (CD0-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40 Ligand, His Tag (Cat. No. CDL-H5248) at 5 μg/mL (100 μL/well) can bind Human CD40, Fc Tag (HPLC-verified) (Cat. No. CD0-H5253) with a linear range of 0.039-1.25 μg/mL (QC tested).;FACS analysis shows that Human CD40, Fc Tag (HPLC-verified) (Cat .No. CD0-H5253) can bind to 293T cells overexpressing human CD40L. The concentration of Human CD40 is 0.1 μg/mL (Routinely tested).
Catalog No. 50-195-7493