Learn More
ACROBiosystems ACROBiosystems Rhesus macaque CD40 / TNFRSF5 Protein, Fc Tag
Supplier: ACROBiosystems CD0C5259100UG
Recombinant Protein;100UG;Rhesus macaque CD40, Fc Tag (CD0-C5259) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # NP_001252791.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.9 kDa. The protein migrates as 56 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Rhesus macaque CD40, Fc Tag (Cat. No. CD0-C5259) at 2 μg/mL (100 μL/well) can bind Human CD40 Ligand, His Tag (Cat. No. CDL-H5140) with a linear range of 5-50 ng/mL (QC tested).