Learn More
Enzo Life Sciences CD40 (human) monoclonal antibody (G28-5) (biotin conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS1290100
Encompass_Preferred
Clone: G28-5. Isotype: IgG1κ. Immunogen: Human erythrocyte-rosette-negative tonsillar lymphocyte. Species reactivity: Human. Applications: Flow Cytometry, IHC (PS). Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2693